COMH04238068 Combi-Blocks, Inc.

5-[(4-Chloro-3,5-dimethyl-1h-pyrazol-1-yl)methyl]-2-furohydrazide

  • MDL number MFCD02253817
  • Cas Number : 386705-59-5
  • Molecular Weight : 268.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4061
Availability details for
Quantity
  • Availability
  • Add to Cart