COMH04238070 Combi-Blocks, Inc.

[(4,8,8-Trimethyl-2-oxo-9,10-dihydro-2h,8h-pyrano[2,3-f]chromen-5-yl)oxy]acetic acid

  • MDL number MFCD06492540
  • Cas Number : 888027-28-9
  • Molecular Weight : 318.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4069
Availability details for
Quantity
  • Availability
  • Add to Cart