COMH042380B9 Combi-Blocks, Inc.

Ethyl (2-methylpiperidin-1-yl)acetate

  • MDL number MFCD06155405
  • Cas Number : 895869-57-5
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4146
Availability details for
Quantity
  • Availability
  • Add to Cart