COMH04238124 Combi-Blocks, Inc.

Ethyl (6-amino-1,3-benzodioxol-5-yl)carbamate

  • MDL number MFCD09743352
  • Cas Number : 936074-68-9
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4255
Availability details for
Quantity
  • Availability
  • Add to Cart