COMH0423812A Combi-Blocks, Inc.

2-[(4-Chloro-2-methylphenyl)amino]-5-nitrobenzonitrile

  • MDL number MFCD09455418
  • Cas Number : 945367-47-5
  • Molecular Weight : 287.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4261
Availability details for
Quantity
  • Availability
  • Add to Cart