COMH04238147 Combi-Blocks, Inc.

(3,4-Dihydro-1h-isochromen-1-ylmethyl)methylamine hydrochloride

  • MDL number MFCD00573693
  • Cas Number : 50683-74-4
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4291
Availability details for
Quantity
  • Availability
  • Add to Cart