COMH0423816E Combi-Blocks, Inc.

Isopropyl 2-amino-5-methyl-4-(4-propylphenyl)thiophene-3-carboxylate

  • MDL number MFCD02090977
  • Cas Number : 905011-49-6
  • Molecular Weight : 317.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4330
Availability details for
Quantity
  • Availability
  • Add to Cart