COMH0423816F Combi-Blocks, Inc.

7-Chloro-2-(3,4-dichlorophenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03422158
  • Cas Number : 863437-98-3
  • Molecular Weight : 366.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4331
Availability details for
Quantity
  • Availability
  • Add to Cart