COMH0423817B Combi-Blocks, Inc.

2-Amino-n-(2,5-dimethoxyphenyl)-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxamide

  • MDL number MFCD03945374
  • Cas Number : 588715-65-5
  • Molecular Weight : 360.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4343
Availability details for
Quantity
  • Availability
  • Add to Cart