COMH042381D3 Combi-Blocks, Inc.

(E)-4-(Cinnamyloxy)benzaldehyde

  • MDL number MFCD06757624
  • Cas Number : 79844-40-9
  • Molecular Weight : 238.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4431
Availability details for
Quantity
  • Availability
  • Add to Cart