COMH042381D7 Combi-Blocks, Inc.

Methyl (4,6,8-trimethyl-2-oxoquinolin-1(2h)-yl)acetate

  • MDL number MFCD07800303
  • Cas Number : 890092-46-3
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4436
Availability details for
Quantity
  • Availability
  • Add to Cart