COMH042381DD Combi-Blocks, Inc.

5-(3,4-Dimethylphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD09266606
  • Cas Number : 848436-25-9
  • Molecular Weight : 335.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4442
Availability details for
Quantity
  • Availability
  • Add to Cart