COMH042381E1 Combi-Blocks, Inc.

2-(Benzylamino)-4-methylpentan-1-ol

  • MDL number MFCD12826138
  • Cas Number : 92111-70-1
  • Molecular Weight : 207.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4446
Availability details for
Quantity
  • Availability
  • Add to Cart