COMH0423822B Combi-Blocks, Inc.

[(3-Hexyl-4,8-dimethyl-2-oxo-2h-chromen-7-yl)oxy]acetic acid

  • MDL number MFCD03016390
  • Cas Number : 438030-13-8
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4522
Availability details for
Quantity
  • Availability
  • Add to Cart