COMH04238242 Combi-Blocks, Inc.

4-(3,4-Dimethoxyphenyl)-4,5,6,7-tetrahydro-3h-imidazo[4,5-c]pyridine

  • MDL number MFCD02115773
  • Cas Number : 46964-25-4
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4545
Availability details for
Quantity
  • Availability
  • Add to Cart