COMH042382BA Combi-Blocks, Inc.

2-Amino-7-(2-thienyl)-7,8-dihydroquinazolin-5(6h)-one

  • MDL number MFCD02634913
  • Cas Number : 424818-72-4
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4666
Availability details for
Quantity
  • Availability
  • Add to Cart