COMH042382CA Combi-Blocks, Inc.

3-(Aminomethyl)-4-hydroxybenzoic acid hydrochloride

  • MDL number MFCD03196947
  • Cas Number : 669703-81-5
  • Molecular Weight : 203.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4682
Availability details for
Quantity
  • Availability
  • Add to Cart