COMH042382D4 Combi-Blocks, Inc.

3-[(4-Chlorophenyl)amino]cyclohex-2-en-1-one

  • MDL number MFCD00129271
  • Cas Number : 36646-75-0
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4692

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4692
Availability details for
Quantity
  • Availability
  • Add to Cart