COMH042382D5 Combi-Blocks, Inc.

1-Acetyl-n-cycloheptylpiperidin-4-amine

  • MDL number MFCD01652803
  • Cas Number : 415947-24-9
  • Molecular Weight : 238.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4693
Availability details for
Quantity
  • Availability
  • Add to Cart