COMH04238302 Combi-Blocks, Inc.

1-(2-Methylcyclohexyl)-1h-benzimidazole-5-carboxylic acid

  • MDL number MFCD03830186
  • Cas Number : 903330-28-9
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4739
Availability details for
Quantity
  • Availability
  • Add to Cart