COMH0423830F Combi-Blocks, Inc.

3-(3,5-Dimethyl-1h-1,2,4-triazol-1-yl)propanenitrile

  • MDL number MFCD14630742
  • Cas Number : 60866-60-6
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4752
Availability details for
Quantity
  • Availability
  • Add to Cart