COMH04238316 Combi-Blocks, Inc.

N,N'-Biphenyl-4,4'-diylbis(2-chloropropanamide)

  • MDL number MFCD07535075
  • Cas Number : 882321-61-1
  • Molecular Weight : 365.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4760
Availability details for
Quantity
  • Availability
  • Add to Cart