COMH04238330 Combi-Blocks, Inc.

Methyl 2-[(cyanoacetyl)amino]-5-isopropylthiophene-3-carboxylate

  • MDL number MFCD03759393
  • Cas Number : 515861-09-3
  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4786
Availability details for
Quantity
  • Availability
  • Add to Cart