COMH04238337 Combi-Blocks, Inc.

2-Amino-n-cyclopropyl-6-(1,1-dimethylpropyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD03422658
  • Cas Number : 590352-39-9
  • Molecular Weight : 306.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4793
Availability details for
Quantity
  • Availability
  • Add to Cart