COMH04238379 Combi-Blocks, Inc.

(4-Oxo-1,4-dihydropyrimido[1,2-a]benzimidazol-2-yl)acetic acid

  • MDL number MFCD11057929
  • Cas Number : 940980-97-2
  • Molecular Weight : 243.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4859

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4859
Availability details for
Quantity
  • Availability
  • Add to Cart