COMH0423838B Combi-Blocks, Inc.

4-(4-Methoxyphenyl)piperazine-1-carboximidamide sulfate

  • MDL number MFCD22421998
  • Cas Number : 1417567-67-9
  • Molecular Weight : 332.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4877
Availability details for
Quantity
  • Availability
  • Add to Cart