COMH042383A3 Combi-Blocks, Inc.

1,3,5-Trimethyl-1h-indole-2-carboxylic acid

  • MDL number MFCD03011748
  • Cas Number : 446830-79-1
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4901
Availability details for
Quantity
  • Availability
  • Add to Cart