COMH042383B6 Combi-Blocks, Inc.

6-Acetyl-2h-1,4-benzothiazin-3(4h)-one

  • MDL number MFCD08446936
  • Cas Number : 91119-38-9
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4920
Availability details for
Quantity
  • Availability
  • Add to Cart