COMH042383C1 Combi-Blocks, Inc.

7-Chloro-2-(2,5-dimethoxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03422156
  • Cas Number : 847503-17-7
  • Molecular Weight : 357.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4931
Availability details for
Quantity
  • Availability
  • Add to Cart