COMH042383E5 Combi-Blocks, Inc.

4-Amino-5-(4-methoxybenzyl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD00698741
  • Cas Number : 93073-14-4
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-4968
Availability details for
Quantity
  • Availability
  • Add to Cart