COMH0423840E Combi-Blocks, Inc.

1-(6-Amino-1,3-benzodioxol-5-yl)pyrrolidin-2-one

  • MDL number MFCD10037589
  • Cas Number : 948007-47-4
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5010
Availability details for
Quantity
  • Availability
  • Add to Cart