COMH0423842D Combi-Blocks, Inc.

(4-Piperidin-1-ylbenzylidene)malononitrile

  • MDL number MFCD00129634
  • Cas Number : 66883-91-8
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5041
Availability details for
Quantity
  • Availability
  • Add to Cart