COMH0423843F Combi-Blocks, Inc.

3-[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD00098187
  • Cas Number : 94192-18-4
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5060
Availability details for
Quantity
  • Availability
  • Add to Cart