COMH04238451 Combi-Blocks, Inc.

Acridin-4-amine hydrochloride

  • MDL number MFCD27756540
  • Cas Number : 578-07-4
  • Molecular Weight : 230.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5078
Availability details for
Quantity
  • Availability
  • Add to Cart