COMH0423846D Combi-Blocks, Inc.

4-(1,3-Dimethyl-2,6-dioxo-2,3,6,9-tetrahydro-1h-purin-8-yl)benzenesulfonic acid

  • MDL number MFCD00545717
  • Cas Number : 80206-91-3
  • Molecular Weight : 336.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5106
Availability details for
Quantity
  • Availability
  • Add to Cart