COMH042384A5 Combi-Blocks, Inc.

1-Acetyl-n-(4-methoxybenzyl)piperidin-4-amine

  • MDL number MFCD01652898
  • Cas Number : 415951-99-4
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5162
Availability details for
Quantity
  • Availability
  • Add to Cart