COMH042384AC Combi-Blocks, Inc.

6-Bromo-2-(4-fluorophenyl)quinoline-4-carboxylic acid

  • MDL number MFCD02056374
  • Cas Number : 391-23-1
  • Molecular Weight : 346.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5169
Availability details for
Quantity
  • Availability
  • Add to Cart