COMH042384B6 Combi-Blocks, Inc.

[1-(([(4-Methylphenyl)sulfonyl]amino)methyl)cyclohexyl]acetic acid

  • MDL number MFCD10007615
  • Cas Number : 675624-80-3
  • Molecular Weight : 325.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5179
Availability details for
Quantity
  • Availability
  • Add to Cart