COMH04238526 Combi-Blocks, Inc.

2-(2,4-Dimethylphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD03421048
  • Cas Number : 883526-18-9
  • Molecular Weight : 295.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5291
Availability details for
Quantity
  • Availability
  • Add to Cart