COMH04238531 Combi-Blocks, Inc.

[3-Chloro-4-(4-propionylpiperazin-1-yl)phenyl]amine

  • MDL number MFCD05843346
  • Cas Number : 838613-75-5
  • Molecular Weight : 267.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5302
Availability details for
Quantity
  • Availability
  • Add to Cart