COMH0423853E Combi-Blocks, Inc.

2-(4-Nitrophenyl)imidazo[2,1-a]isoquinoline

  • MDL number MFCD01721897
  • Cas Number : 61001-13-6
  • Molecular Weight : 289.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5315
Availability details for
Quantity
  • Availability
  • Add to Cart