COMH0423859D Combi-Blocks, Inc.

2-(2,4-Difluorophenoxy)-2-methylpropanoyl chloride

  • MDL number MFCD12197856
  • Cas Number : 936327-47-8
  • Molecular Weight : 234.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5411
Availability details for
Quantity
  • Availability
  • Add to Cart