COMH042385D4 Combi-Blocks, Inc.

2-([4-(Methoxycarbonyl)phenyl]amino)-2-oxoethyl 4-aminobenzoate

  • MDL number MFCD09669461
  • Cas Number : 931239-21-3
  • Molecular Weight : 328.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5467
Availability details for
Quantity
  • Availability
  • Add to Cart