COMH042385EE Combi-Blocks, Inc.

3-(4-Hydroxyphenyl)adamantane-1-carboxylic acid

  • MDL number MFCD01678008
  • Cas Number : 56531-55-6
  • Molecular Weight : 272.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5493
Availability details for
Quantity
  • Availability
  • Add to Cart