COMH042385F7 Combi-Blocks, Inc.

2-[(2-Methoxybenzyl)amino]-5-nitrobenzonitrile

  • MDL number MFCD04690404
  • Cas Number : 785798-40-5
  • Molecular Weight : 283.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5502
Availability details for
Quantity
  • Availability
  • Add to Cart