COMH04238611 Combi-Blocks, Inc.

5-Furan-2-ylmethylene-thiazolidine-2,4-dione

  • MDL number MFCD00460127
  • Cas Number : 5718-88-7
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5528
Availability details for
Quantity
  • Availability
  • Add to Cart