COMH04238638 Combi-Blocks, Inc.

8-Chloro-2-(3,4-dimethoxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD03422153
  • Cas Number : 862782-46-5
  • Molecular Weight : 343.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5567

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5567
Availability details for
Quantity
  • Availability
  • Add to Cart