COMH0423864A Combi-Blocks, Inc.

Ethyl 2-amino-4-(3,4-dichlorophenyl)thiophene-3-carboxylate

  • MDL number MFCD01923246
  • Cas Number : 351156-69-9
  • Molecular Weight : 316.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5585
Availability details for
Quantity
  • Availability
  • Add to Cart