COMH0423865C Combi-Blocks, Inc.

3-Chloro-4-(2-naphthyloxy)aniline

  • MDL number MFCD00437172
  • Cas Number : 71311-87-0
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5603
Availability details for
Quantity
  • Availability
  • Add to Cart