COMH04238675 Combi-Blocks, Inc.

5-Chloro-3-[n-(methoxy-carbonyl-methyl)sulfamoyl]-2-thiophene carboxylic acid methyl ester

  • MDL number MFCD09455046
  • Cas Number : 906522-87-0
  • Molecular Weight : 327.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-5629
Availability details for
Quantity
  • Availability
  • Add to Cart